Methyl 2-chloro-6-piperidin-1-ylisonicotinate

Lieferant: Apollo Scientific

APOSOR110253-1GEA 213 EUR
APOSOR110253-1G
Methyl 2-chloro-6-piperidin-1-ylisonicotinate
Methyl 2-chloro-6-piperidin-1-ylisonicotinate
Formel: C₁₂H₁₅ClN₂O₂
Molekulargewicht: 254,713 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12545989
CAS-Nummer: 1227955-17-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR