3-Chloroimidazo[1,2-a]pyridine
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR110406-1GEA
81.5
EUR
APOSOR110406-1G
3-Chloroimidazo[1,2-a]pyridine
3-Chloroimidazo[1,2-a]pyridine
Formel:
C₇H₅ClN₂ Molekulargewicht: 152,581 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD07778355 CAS-Nummer: 5315-73-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...