1H-Benzimidazole-5-carbohydrazide
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR110435-1GEA
234
EUR
APOSOR110435-1G
1H-Benzimidazole-5-carbohydrazide
1H-Benzimidazole-5-carbohydrazide
Formel:
C₈H₈N₄O Molekulargewicht: 176,175 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08703627 CAS-Nummer: 108038-52-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...