1-Phenylcyclobutanecarboxamide

Lieferant: Apollo Scientific

APOSOR110488-1GEA 222 EUR
APOSOR110488-1G
1-Phenylcyclobutanecarboxamide
1-Phenylcyclobutanecarboxamide
Formel: C₁₁H₁₃NO
Molekulargewicht: 175,227 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD10664451
CAS-Nummer: 17380-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR