2-(4-Isopropylphenoxy)benzaldehyde

APOSOR110500-1GEA 0 EUR
APOSOR110500-1G APOSOR110500-5G
2-(4-Isopropylphenoxy)benzaldehyde
2-(4-Isopropylphenoxy)benzaldehyde
Formel: C₁₆H₁₆O₂
Molekulargewicht: 240,297 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11521736
CAS-Nummer: 909789-04-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR