1-[4-(Bromomethyl)phenoxy]-3-methylbenzene

Lieferant: Apollo Scientific

APOSOR110527-1GEA 350 EUR
APOSOR110527-1G
1-[4-(Bromomethyl)phenoxy]-3-methylbenzene
1-[4-(Bromomethyl)phenoxy]-3-methylbenzene
Formel: C₁₄H₁₃BrO
Molekulargewicht: 277,156 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD16619598
CAS-Nummer: 95922-53-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR