1-(4-Bromophenyl)cyclobutane-1-carbonitrile

Lieferant: Apollo Scientific
Achtung

APOSOR110546-1GEA 292 EUR
APOSOR110546-1G
1-(4-Bromophenyl)cyclobutane-1-carbonitrile
1-(4-Bromophenyl)cyclobutane-1-carbonitrile
Formel: C₁₁H₁₀BrN
Molekulargewicht: 236,108 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD10688823
CAS-Nummer: 485828-58-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR