4-(3-Ethyl-1,2,4-oxadiazol-5-yl)benzamide

Lieferant: Apollo Scientific

APOSOR110549-1GEA 373 EUR
APOSOR110549-1G
4-(3-Ethyl-1,2,4-oxadiazol-5-yl)benzamide
4-(3-Ethyl-1,2,4-oxadiazol-5-yl)benzamide
Formel: C₁₁H₁₁N₃O₂
Molekulargewicht: 217,224 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD18384923
CAS-Nummer: 1427460-93-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR