Methyl 4-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxylate

Lieferant: Apollo Scientific
Achtung

APOSOR110752-1GEA 350 EUR
APOSOR110752-1G
Methyl 4-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxylate
Methyl 4-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxylate
Formel: C₁₁H₁₁NO₄
Molekulargewicht: 221,209 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12756248
CAS-Nummer: 1017273-27-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR