5-(4-tert-Butylphenyl)-1-phenyl-1,2-dihydro-3H-1,2,4-triazole-3-thione
Lieferant: Apollo Scientific
APOSOR110850-1GEA
350
EUR
APOSOR110850-1G
5-(4-tert-Butylphenyl)-1-phenyl-1,2-dihydro-3H-1,2,4-triazole-3-thione
5-(4-tert-Butylphenyl)-1-phenyl-1,2-dihydro-3H-1,2,4-triazole-3-thione
Formel:
C₁₈H₁₉N₃S Molekulargewicht: 309,429 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD26936321 CAS-Nummer: 1858255-56-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...