1-(5-Bromo-1-benzofuran-2-yl)-3-(dimethylamino)prop-2-en-1-one

Lieferant: Apollo Scientific

APOSOR110916-1GEA 292 EUR
APOSOR110916-1G
1-(5-Bromo-1-benzofuran-2-yl)-3-(dimethylamino)prop-2-en-1-one
1-(5-Bromo-1-benzofuran-2-yl)-3-(dimethylamino)prop-2-en-1-one
Formel: C₁₃H₁₂BrNO₂
Molekulargewicht: 294,144 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD19000256
CAS-Nummer: 1563069-87-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR