Thieno[2,3-b]pyridine-2-carboxamide

Lieferant: Apollo Scientific

APOSOR110964-1GEA 302 EUR
APOSOR110964-1G
Thieno[2,3-b]pyridine-2-carboxamide
Thieno[2,3-b]pyridine-2-carboxamide
Formel: C₈H₆N₂OS
Molekulargewicht: 178,211 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD28103331
CAS-Nummer: 1824270-94-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR