2-Isocyanato-4,5,6,7-tetrahydro-1-benzothiophene

Lieferant: Apollo Scientific

APOSOR111339-1GEA 339 EUR
APOSOR111339-1G
2-Isocyanato-4,5,6,7-tetrahydro-1-benzothiophene
2-Isocyanato-4,5,6,7-tetrahydro-1-benzothiophene

May form small amount of sediment

Formel: C₉H₉NOS
Molekulargewicht: 179,239 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11130324
CAS-Nummer: 1019374-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR