2-Acetamidothiophene-3-carboxylic acid

Lieferant: Apollo Scientific

APOSOR111371-5GEA 789 EUR
APOSOR111371-5G APOSOR111371-10G APOSOR111371-1G
2-Acetamidothiophene-3-carboxylic acid
2-Acetamidothiophene-3-carboxylic acid
Formel: C₇H₇NO₃S
Molekulargewicht: 185,2 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00159566
CAS-Nummer: 51419-38-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR