Diethyl 4-bromophenylmalonate
APOSOR111375-5GEA
0
EUR
APOSOR111375-5G
APOSOR111375-1G
Diethyl 4-bromophenylmalonate
Diethyl 4-bromophenylmalonate
Formel:
C₁₃H₁₅BrO₄ Molekulargewicht: 315,16 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06205062 CAS-Nummer: 93139-85-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...