3-Amino-5-chloro-1-benzofuran-2-carboxylic acid

Lieferant: Apollo Scientific

APOSOR111414-5GEA 677 EUR
APOSOR111414-5G APOSOR111414-1G
3-Amino-5-chloro-1-benzofuran-2-carboxylic acid
3-Amino-5-chloro-1-benzofuran-2-carboxylic acid
Formel: C₉H₆ClNO₃
Molekulargewicht: 211,602 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD22988712
CAS-Nummer: 894796-29-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR