Ethyl (4-amino-3-bromophenyl)acetate

Lieferant: Apollo Scientific

APOSOR111420-1GEA 282 EUR
APOSOR111420-1G
Ethyl (4-amino-3-bromophenyl)acetate
Ethyl (4-amino-3-bromophenyl)acetate
Formel: C₁₀H₁₂BrNO₂
Molekulargewicht: 258,112 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12547819
CAS-Nummer: 152362-31-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR