[4-(2-Oxoimidazolidin-1-yl)phenyl]acetic acid

Lieferant: Apollo Scientific

APOSOR111422-1GEA 517 EUR
APOSOR111422-1G
[4-(2-Oxoimidazolidin-1-yl)phenyl]acetic acid
[4-(2-Oxoimidazolidin-1-yl)phenyl]acetic acid
Formel: C₁₁H₁₂N₂O₃
Molekulargewicht: 220,225 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD05864552
CAS-Nummer: 492445-92-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR