5-Methoxy-1-methyl-3-(1,3-oxazol-5-yl)-1H-indole
APOSOR111560-1GEA
0
EUR
APOSOR111560-1G
5-Methoxy-1-methyl-3-(1,3-oxazol-5-yl)-1H-indole
5-Methoxy-1-methyl-3-(1,3-oxazol-5-yl)-1H-indole
Formel:
C₁₃H₁₂N₂O₂ Molekulargewicht: 228,247 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD31629935 CAS-Nummer: 2244087-86-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...