2-Oxochromane-6-sulphonyl chloride

Lieferant: Apollo Scientific

APOSOR111583-1GEA 234 EUR
APOSOR111583-1G
2-Oxochromane-6-sulphonyl chloride
2-Oxochromane-6-sulphonyl chloride
Formel: C₉H₇ClO₄S
Molekulargewicht: 246,667 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11054178
CAS-Nummer: 600166-90-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR