1-(5-Chloro-2-hydroxyphenyl)ethanone (2,4,6-trichlorophenyl)hydrazone

Lieferant: Apollo Scientific

APOSOR111587-1GEA 466 EUR
APOSOR111587-1G
1-(5-Chloro-2-hydroxyphenyl)ethanone (2,4,6-trichlorophenyl)hydrazone
1-(5-Chloro-2-hydroxyphenyl)ethanone (2,4,6-trichlorophenyl)hydrazone
Formel: C₁₄H₁₀Cl₄N₂O
Molekulargewicht: 364,054 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00116814
CAS-Nummer: 219315-22-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR