3-[(4-Bromophenyl)carbamoyl]benzeneboronic acid 95%

Lieferant: Apollo Scientific

APOSOR11448-5GEA 401 EUR
APOSOR11448-5G APOSOR11448-1G
3-[(4-Bromophenyl)carbamoyl]benzeneboronic acid 95%
3-[(4-Bromophenyl)carbamoyl]benzeneboronic acid
Formel: C₁₃H₁₁BBrNO₃
Molekulargewicht: 319,946 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09475817
CAS-Nummer: 874288-28-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR