4-(3-Morpholin-4-ylpropoxy)benzeneboronic acid, pinacol ester 98%

Achtung

APOSOR11457-250MGEA 0 EUR
APOSOR11457-250MG APOSOR11457-1G
4-(3-Morpholin-4-ylpropoxy)benzeneboronic acid, pinacol ester 98%
4-(3-Morpholin-4-ylpropoxy)benzeneboronic acid, pinacol ester
Formel: C₁₉H₃₀BNO₄
Molekulargewicht: 347,257 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04440858
CAS-Nummer: 910462-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR