1-[(2-Bromophenyl)sulphonyl]-3,5-dimethyl-1H-pyrazole

APOSOR11576-5GEA 0 EUR
APOSOR11576-5G APOSOR11576-1G
1-[(2-Bromophenyl)sulphonyl]-3,5-dimethyl-1H-pyrazole
1-[(2-Bromophenyl)sulphonyl]-3,5-dimethyl-1H-pyrazole
Formel: C₁₁H₁₁BrN₂O₂S
Molekulargewicht: 315,186 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09800985
CAS-Nummer: 957120-77-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR