2',7'-Dibromospiro[cyclopent[3]ene-1,9'-fluorene]

Lieferant: Apollo Scientific
Achtung

APOSOR11618-1GEA 270 EUR
APOSOR11618-1G APOSOR11618-5G
2',7'-Dibromospiro[cyclopent[3]ene-1,9'-fluorene]
2',7'-Dibromospiro[cyclopent[3]ene-1,9'-fluorene]
Formel: C₁₇H₁₂Br₂
Molekulargewicht: 376,085 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09800925
CAS-Nummer: 951884-04-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR