5-(Methoxycarbonyl)thiophene-3-boronic acid 96%
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR11695-1GEA
343
EUR
APOSOR11695-1G
5-(Methoxycarbonyl)thiophene-3-boronic acid 96%
5-(Methoxycarbonyl)thiophene-3-boronic acid
Formel:
C₆H₇BO₄S Molekulargewicht: 185,993 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD09800878 CAS-Nummer: 957062-52-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...