4-(2-Chloroethyl)piperazine hydrochloride, N1-BOC protected 98%

Lieferant: Apollo Scientific

APOSOR11730-25GEA 637 EUR
APOSOR11730-25G APOSOR11730-10G APOSOR11730-5G
4-(2-Chloroethyl)piperazine hydrochloride, N1-BOC protected 98%
4-(2-Chloroethyl)piperazine hydrochloride, N1-BOC protected
Formel: C₁₁H₂₁ClN₂O₂·ClH
Molekulargewicht: 285,211 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09800597
CAS-Nummer: 1170890-35-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR