4-Chloro-6-[(3-methoxypropyl)amino]-2-(methylthio)pyrimidine
APOSOR11960-1GEA
0
EUR
APOSOR11960-1G
4-Chloro-6-[(3-methoxypropyl)amino]-2-(methylthio)pyrimidine
4-Chloro-6-[(3-methoxypropyl)amino]-2-(methylthio)pyrimidine
Formel:
C₉H₁₄ClN₃OS Molekulargewicht: 247,745 g/mol Schmelzpunkt: 60…64 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD09800996 CAS-Nummer: 951884-56-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...