4-Bromo-1-butyl-1H-pyrazole
Lieferant: Apollo Scientific
APOSOR11984-1GEA
102
EUR
APOSOR11984-1G
APOSOR11984-5G
4-Bromo-1-butyl-1H-pyrazole
4-Bromo-1-butyl-1H-pyrazole
Formel:
C₇H₁₁BrN₂ Molekulargewicht: 203,08 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09801022 CAS-Nummer: 957062-61-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...