3-(2-Methyl-1H-imidazol-1-yl)benzoic acid 90%
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR12218-250MGEA
149
EUR
APOSOR12218-250MG
APOSOR12218-1G
3-(2-Methyl-1H-imidazol-1-yl)benzoic acid 90%
3-(2-Methyl-1H-imidazol-1-yl)benzoic acid
Formel:
C₁₁H₁₀N₂O₂ Molekulargewicht: 202,209 g/mol Schmelzpunkt: 182,5…184.5 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09065014 CAS-Nummer: 898289-59-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...