2-[(Methylamino)methyl]pyridine dihydrochloride

Gefahr

APOSOR12501-1GEA 0 EUR
APOSOR12501-1G APOSOR12501-5G
2-[(Methylamino)methyl]pyridine dihydrochloride
2-[(Methylamino)methyl]pyridine dihydrochloride
Formel: C₇H₁₀N₂·2ClH
Molekulargewicht: 195,09 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD03840005
CAS-Nummer: 100868-72-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR