4-(Aminomethyl)-4'-chlorobiphenyl hydrochloride

Lieferant: Apollo Scientific
Achtung

APOSOR12633-100MGEA 222 EUR
APOSOR12633-100MG APOSOR12633-1G APOSOR12633-250MG
4-(Aminomethyl)-4'-chlorobiphenyl hydrochloride
4-(Aminomethyl)-4'-chlorobiphenyl hydrochloride
Formel: C₁₃H₁₂ClN·ClH
Molekulargewicht: 254,155 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD02089424
CAS-Nummer: 410077-96-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR