3-(Pyridin-3-yl)morpholine

Achtung

APOSOR12858-250MGEA 0 EUR
APOSOR12858-250MG
3-(Pyridin-3-yl)morpholine
3-(Pyridin-3-yl)morpholine
Formel: C₉H₁₂N₂O
Molekulargewicht: 164,204 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD04113629
CAS-Nummer: 887344-25-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR