3-[(Pyrrolidin-3-yl)methyl]pyridine oxalate
Lieferant: Apollo Scientific
|
Gefahr
|
APOSOR12929-50MGEA
36.4
EUR
APOSOR12929-50MG
3-[(Pyrrolidin-3-yl)methyl]pyridine oxalate
3-[(Pyrrolidin-3-yl)methyl]pyridine oxalate
Formel:
C₁₀H₁₄N₂·C₂H₂O₄ Molekulargewicht: 252,266 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD04117738 CAS-Nummer: 1018827-46-7 UN-Nummer: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...