3,5-Dimethyl-4-formyl-1H-pyrazole, N1-BOC protected
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR1389-250MGEA
132
EUR
APOSOR1389-250MG
APOSOR1389-1G
3,5-Dimethyl-4-formyl-1H-pyrazole, N1-BOC protected
3,5-Dimethyl-4-formyl-1H-pyrazole, N1-BOC protected
Formel:
C₁₁H₁₆N₂O₃ Molekulargewicht: 224,256 g/mol Schmelzpunkt: 59…62 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06659076 CAS-Nummer: 844891-13-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...