N-(1,3-Benzodioxol-5-yl)glycinehydrazide

Lieferant: Apollo Scientific
Achtung

APOSOR14397-1GEA 270 EUR
APOSOR14397-1G
N-(1,3-Benzodioxol-5-yl)glycinehydrazide
N-(1,3-Benzodioxol-5-yl)glycinehydrazide
Formel: C₉H₁₁N₃O₃
Molekulargewicht: 209,202 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00098841
CAS-Nummer: 271798-51-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR