3-Chloro-6,7-dihydro-5H-benzo[6,7]cyclohepta[1,2-c]pyridazine
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR14956-250MGEA
742
EUR
APOSOR14956-250MG
3-Chloro-6,7-dihydro-5H-benzo[6,7]cyclohepta[1,2-c]pyridazine
3-Chloro-6,7-dihydro-5H-benzo[6,7]cyclohepta[1,2-c]pyridazine
Formel:
C₁₃H₁₁ClN₂ Molekulargewicht: 230,693 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06801335 CAS-Nummer: 25823-53-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...