2-(4-Nitrophenyl)cyclooctan-1-one

Lieferant: Apollo Scientific

APOSOR340129-100MGEA 626 EUR
APOSOR340129-100MG
2-(4-Nitrophenyl)cyclooctan-1-one
2-(4-Nitrophenyl)cyclooctan-1-one
Formel: C₁₄H₁₇NO₃
Molekulargewicht: 247,29 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD28154742
CAS-Nummer: 1451450-77-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR