Methyl 2-(4-nitronapthalen-1-yl)-2-phenylacetate

APOSOR340140-100MGEA 0 EUR
APOSOR340140-100MG
Methyl 2-(4-nitronapthalen-1-yl)-2-phenylacetate
Methyl 2-(4-nitronapthalen-1-yl)-2-phenylacetate
Formel: C₁₉H₁₅NO₄
Molekulargewicht: 321,327 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD28154753
CAS-Nummer: 1980086-65-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR