3-(4-Nitrophenyl)-indolin-2-one

APOSOR340154-100MGEA 0 EUR
APOSOR340154-100MG
3-(4-Nitrophenyl)-indolin-2-one
3-(4-Nitrophenyl)-indolin-2-one
Formel: C₁₄H₁₀N₂O₃
Molekulargewicht: 254,241 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD28154767
CAS-Nummer: 53497-60-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR