2-(p-Tolyl)benzofuran

APOSOR340160-100MGEA 0 EUR
APOSOR340160-100MG
2-(p-Tolyl)benzofuran
2-(p-Tolyl)benzofuran
Formel: C₁₅H₁₂O
Molekulargewicht: 208,255 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD22989384
CAS-Nummer: 25664-48-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR