2-(2,5-Dichlorophenyl)-benzofuran

APOSOR340165-100MGEA 0 EUR
APOSOR340165-100MG
2-(2,5-Dichlorophenyl)-benzofuran
2-(2,5-Dichlorophenyl)-benzofuran
Formel: C₁₄H₈Cl₂O
Molekulargewicht: 263,119 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD28154773
CAS-Nummer: 65246-46-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR