2-(2-((tert-Butyldimethylsilyl)oxy)ethyl)-3-ethylaziridine

Lieferant: Apollo Scientific

APOSOR340178-100MGEA 626 EUR
APOSOR340178-100MG
2-(2-((tert-Butyldimethylsilyl)oxy)ethyl)-3-ethylaziridine
2-(2-((tert-Butyldimethylsilyl)oxy)ethyl)-3-ethylaziridine
Formel: C₁₂H₂₇NOSi
Molekulargewicht: 229,434 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD28154782
CAS-Nummer: 1534354-34-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR