2-Hydroxy-3,5-dibromo-4-methylpyridine 95+%

Lieferant: Apollo Scientific

APOSOR345147-1GEA 104 EUR
APOSOR345147-1G
2-Hydroxy-3,5-dibromo-4-methylpyridine 95+%
2-Hydroxy-3,5-dibromo-4-methylpyridine
Formel: C₆H₅Br₂NO
Molekulargewicht: 266,918 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06254604
CAS-Nummer: 89581-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR