2-Chloro-5-methoxypyridine 98%

APOSOR345253-1GEA 0 EUR
APOSOR345253-1G APOSOR345253-5G
2-Chloro-5-methoxypyridine 98%
2-Chloro-5-methoxypyridine
Formel: C₆H₆ClNO
Molekulargewicht: 143,571 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06659513
CAS-Nummer: 139585-48-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR