3-Bromo-5-chloro-2-iodopyridine 97%

Lieferant: Apollo Scientific

APOSOR345417-250MGEA 80 EUR
APOSOR345417-250MG
3-Bromo-5-chloro-2-iodopyridine 97%
3-Bromo-5-chloro-2-iodopyridine
Formel: C₅H₂BrClIN
Molekulargewicht: 318,338 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11847354
CAS-Nummer: 823221-97-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR