4-Chloro-6-methyl-2-(methylthio)pyrimidine-5-carbonitrile

Lieferant: Apollo Scientific

APOSOR345713-1GEA 187 EUR
APOSOR345713-1G
4-Chloro-6-methyl-2-(methylthio)pyrimidine-5-carbonitrile
4-Chloro-6-methyl-2-(methylthio)pyrimidine-5-carbonitrile
Formel: C₇H₆ClN₃S
Molekulargewicht: 199,661 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD28134480
CAS-Nummer: 1208170-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR