Ethyl 2-(2-bromobenzyl)thiazole-4-carboxylate

Lieferant: Apollo Scientific

APOSOR346068-1GEA 317 EUR
APOSOR346068-1G
Ethyl 2-(2-bromobenzyl)thiazole-4-carboxylate
Ethyl 2-(2-bromobenzyl)thiazole-4-carboxylate
Formel: C₁₃H₁₂BrNO₂S
Molekulargewicht: 326,209 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD20526238
CAS-Nummer: 1351479-04-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR