4-[4-(1,3-Dioxo-1,3-dihydro-isoindol-2-yl)-butyl]-piperazine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

APOSOR346283-1GEA 286 EUR
APOSOR346283-1G
4-[4-(1,3-Dioxo-1,3-dihydro-isoindol-2-yl)-butyl]-piperazine-1-carboxylic acid tert-butyl ester
4-[4-(1,3-Dioxo-1,3-dihydro-isoindol-2-yl)-butyl]-piperazine-1-carboxylic acid tert-butyl ester
Formel: C₂₁H₂₉N₃O₄
Molekulargewicht: 387,473 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD19981496
CAS-Nummer: 745048-06-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR