3-Methyl-isothiazole-5-sulphonyl chloride

Lieferant: Apollo Scientific

APOSOR346292-1GEA 412 EUR
APOSOR346292-1G APOSOR346292-500MG
3-Methyl-isothiazole-5-sulphonyl chloride
3-Methyl-isothiazole-5-sulphonyl chloride
Formel: C₄H₄ClNO₂S₂
Molekulargewicht: 197,663 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD21362487
CAS-Nummer: 1355334-86-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR